Fisherbrand™ Kahn Serological Pipets
For measuring and transferring serum and antigen in Kahn tests.
$282.03 - $341.45
Specifications
Material | Borosilicate Glass |
---|---|
Graduated | Yes |
Graduations | 0.001 mL |
Type | Serological Pipet |
Description
Used in Kahn tests as published in American Journal of Public Health, 129 (June 1924).
- Accuracy printed on each pipet
- Borosilicate glass with permanent amber markings
- Tuf-Temp™ treated to reduce chipping
Compliance
Calibrated “To Deliver/Blow Out.” Pipet 13-677D meets ASTM™ E 1044-96.
Specifications
Borosilicate Glass | |
Yes | |
0.001 mL | |
Serological Pipet |