Please login to your online account to display your discounted pricing
Fisherbrand™ Intermediate-Form Polypropylene Beakers with Handles
Chemical-resistant, with a convenient pouring spout
$24.43 - $83.54
Specifications
Autoclavable Yes
Material PP (Polypropylene)
Product Type Beaker
For Use With (Application) Chemical-resistant, with a convenient pouring spout
Graduated Yes
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) Price Quantity    
02-591-36
  Accessories
fisherbrand™
5264750050F
50mL Pack of 5 Each for $80.50
02-591-37
  Accessories
fisherbrand™
5264750100F
100mL Pack of 5 Each for $83.54
02-591-38
  Accessories
fisherbrand™
5264750250F
250mL Pack for $24.43
02-591-39
fisherbrand™
5264750500F
500mL Pack for $26.79
Description & Specifications
Specifications
Autoclavable Yes
Material PP (Polypropylene)
Product Type Beaker
For Use With (Application) Chemical-resistant, with a convenient pouring spout
Graduated Yes
Includes Handle, Spout
Scale Blue graduated scale
Temperature (English) Max. Operating 250°F
Temperature (Metric) Max. Operating 121°C
  • With ergonomic, comfortable handles for safer pouring
  • Beaker bottoms are designed so they will not slide or create suction and stick to laboratory countertops
  • Graduations in liters and milliliters are in easy-to-read blue print
  • Autoclavable