Please login to your online account to display your discounted pricing

Fisherbrand™ Industrial-Grade Vinyl Cleanroom Tape

For light industrial use, clean construction and cleanroom support

$8.57 - $474.46

Specifications

Certifications/Compliance FDA, CID
Format Tape
Product Type Industrial grade cleanroom Tape
Quantity Each
Sterile No
View More Specs
Products
Catalog Number Mfr. No. Width (English) Color Price Quantity    

19-095-104

 
fisherbrand™
1WH-EGVFP
1 in. White Case of 36 Each for $474.46

19-120-3204

 
fisherbrand™
EGVFP1LB
1 in. Light Blue Each for $12.41
Case of 36 Each for $422.28

19120-3203

 
fisherbrand™
EGVFP1DG
1 in. Dark Green Each for $12.41
Case of 36 Each for $422.28

19120-3209

 
fisherbrand™
EGVFP12BL
0.5 in. Black Each for $8.57
Case of 72 Each for $583.20

19120-3214

 
fisherbrand™
EGVFP12LG
0.5 in. Light Green Each for $8.81
Case of 72 Each for $599.04

19120-3212

 
fisherbrand™
EGVFP12DG
0.5 in. Dark Green Each for $8.81
Case of 72 Each for $599.04

19120-3213

 
fisherbrand™
EGVFP12LB
0.5 in. Light Blue Each for $8.81
Case of 72 Each for $599.04

19120-3211

 
fisherbrand™
EGVFP12DB
0.5 in. Dark Blue Each for $8.81
Case of 72 Each for $599.04

19120-3215

 
fisherbrand™
EGVFP12OR
0.5 in. Orange Each for $8.81
Case of 72 Each for $599.04

19120-3217

 
fisherbrand™
EGVFP12RE
0.5 in. Red Each for $8.81
Case of 72 Each for $599.04

19120-3216

 
fisherbrand™
EGVFP12PU
0.5 in. Purple Each for $8.81
Case of 72 Each for $599.04

19120-3218

 
fisherbrand™
EGVFP12TR
0.5 in. Transparent Each for $8.81
Case of 72 Each for $599.04

19120-3220

 
fisherbrand™
EGVFP15BL
1.5 in. Black Each for $14.76
Case of 24 Each for $334.80

19120-3219

 
fisherbrand™
EGVFP12YE
0.5 in. Yellow Each for $8.81
Case of 72 Each for $599.04

19120-3221

 
fisherbrand™
EGVFP15BR
1.5 in. Brown Each for $14.76
Case of 24 Each for $334.80

19120-3222

 
fisherbrand™
EGVFP15DB
1.5 in. Dark Blue Each for $14.76
Case of 24 Each for $334.80

19120-3225

 
fisherbrand™
EGVFP15LG
1.5 in. Light Green Each for $14.76
Case of 24 Each for $334.80

19120-3224

 
fisherbrand™
EGVFP15LB
1.5 in. Light Blue Each for $14.76
Case of 24 Each for $334.80

19120-3226

 
fisherbrand™
EGVFP15OR
1.5 in. Orange Each for $14.76
Case of 24 Each for $334.80

19120-3227

 
fisherbrand™
EGVFP15PU
1.5 in. Purple Each for $14.76
Case of 24 Each for $334.80

19120-3228

 
fisherbrand™
EGVFP15RE
1.5 in. Red Each for $14.76
Case of 24 Each for $334.80

19120-3230

 
fisherbrand™
EGVFP15WH
1.5 in. White Each for $14.76
Case of 24 Each for $334.80

19120-3229

 
fisherbrand™
EGVFP15TR
1.5 in. Transparent Each for $14.76
Case of 24 Each for $334.80

19120-3231

 
fisherbrand™
EGVFP15YE
1.5 in. Yellow Each for $14.76
Case of 24 Each for $334.80

19120-3249

 
fisherbrand™
EGVFP34OR
0.75 in. Orange Each for $9.29
Case of 48 Each for $396.96
Description & Specifications

Specifications

Certifications/Compliance FDA, CID
Format Tape
Product Type Industrial grade cleanroom Tape
Quantity Each
Sterile No
For Use With (Application) Aisle marking, labeling, batching, clean-build projects, light industrial use and food processing applications
Grade Industrial
Length (English) 36 yd.
Length (Metric) 33m
Material Vinyl

  • Designed for use in areas where ultraclean tape is not required
  • Forms a strong bond with plastics, polymers and stainless steel without leaving adhesive residue when removed
  • Can be written on with cleanroom pen
  • Processed in a controlled environment
  • Wound on a plastic core and sleeve bagged
  • All rolls are 36 yd. long (33m)

Complies with Federal Specifications PPP-T-66E and CID A-A 1689A