Please login to your online account to display your discounted pricing

Fisherbrand™ Individual Filter Cartridges for Milli-Q™ Pure Water Polishing Systems


High-quality filters fit Millipore™ Milli-Q three- or four-bowl Systems—without expensive hardware or conversion kits

Manufacturer: fisherbrand™  Y2746

 View more versions of this product

Catalog No. 09-025-10

  • / Each
  • / Case of 10 Each
 

Description & Specifications

Specifications

Media Type Acid/Base Nuclear-grade Mixed-Bed
Resin Type Mixed bed
For Use With (Equipment) Milli-Q Pure Water Polishing Systems
Capacity (English) 400 gr. as CaCO2
Product Type Ion exchange filter cartridge
Shelf Life 1 Year
Accessories

1 Year