Fisherbrand™ Individual Filter Cartridges for Milli-Q™ Pure Water Polishing Systems
High-quality filters fit Millipore™ Milli-Q three- or four-bowl Systems—without expensive hardware or conversion kits
Manufacturer:  Fisherbrand™ Y2748
 View more versions of this product
Catalog No. 09-025-17
Specifications Description & Specifications
Specifications
0.25 in.
Milli-Q Pure Water Polishing Systems
0.2μm
0.5
Yes
Hollow Fiber filter cartridge
Polycarbonate