Fisherbrand™ Humidity Sponge™
Turns any container into a desiccator.
$70.20 - $71.55
Specifications
For Use With (Application) Turns any container into a desiccator
Weight (English) 0.02 lb.
Weight (Metric) 10g
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
S33516IND Fisherbrand™
3150EMD
Humidity Sponge Indicating Desiccant Pack for $71.55
S33516REG Fisherbrand™
3151EMD
Humidity Sponge Desiccant Pack for $70.20
Specifications
Description
  • Meets all scientific drying requirements
  • Drying capacity depends on volume
Indicator Pack contains a blue indicating gel that turns pink when the desiccant pack has reached its moisture capacity and must be replaced.
Reusable Pack may be reused repeatedly and does not have a color indicator. Regeneration requires oven heating to 104°C (220°F). Single relative humidity card shows when regeneration is required.
Description & Specifications
Specifications
Turns any container into a desiccator
0.02 lb.
10g