Fisherbrand™ High-Purity Stainless-Steel Single-Stage Regulators
For semi-corrosive gases
Manufacturer:  Genstar Technologies Co 10575127
 View more versions of this product
Catalog No. 10-575-127
Specifications Description & Specifications
Specifications
Stainless Steel
2 to 40psi
3000psi
316L Stainless Steel
0.25 in. NPT outlet port
10μ Sintered
-40 to 140°F
5.08cm
PCTFE
-40 to 60°C
Single
Semi corrosive gases, extremely high purity gases, mildly corrosive or reactive gases, and toxic gases where slight variations in delivery pressure can be tolerated
30 in. Hg 0 to 60psi
2 in.
Gas regulator
PTFE Resin