Please login to your online account to display your discounted pricing

Fisherbrand™ High-Form Alumina Crucibles Covers


Covers for High-Form Alumina crucibles

Manufacturer: fisherbrand™  HAL100

 View more versions of this product

Catalog No. FB9604

  • / Pack of 6 Each
  • / Case of 4 Pack
 

Description & Specifications

Specifications

Product Type Crucible Cover
For Use With (Application) For 100mL crucible
Material Alumina
Color Cream