missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Heavy-Duty Rectangular Cast-Iron Support

For maximum stability and molded polyethylene tray for spillage protection

Manufacturer:  Fisherbrand™ 00576Q

Catalog No. 14-679Q



Description

Description

For maximum stability and molded polyethylene tray for spillage protection

  • Concentric rings on tray provide convenient guide for positioning containers
  • Height to top of rod is 35 in. (89cm)
  • With steel rod and rubber feet to prevent sliding

Specifications

Specifications

Heavy Duty Support
Rectangular
9.75 in.
Each
23cm
Cast Iron
Steel rod and rubber feet to prevent sliding
25cm
9.18 in.
SDS
Documents
Accessories