missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Heavy Duty Polypropylene Lab Trays

Fabricated with 3/16 in. thick polypropylene.

Manufacturer:  Fisherbrand™ 107354

 View more versions of this product

Catalog No. 15-236-1F



Description

Description

Fabricated with 3/16 in. thick polypropylene.

  • Rectangular with welded corners and bottom
  • Can be customized with spigotes and covers
  • Won't rust

Order Info

Additional spill tray sizes available. To order, contact your Fisher Scientific Customer Service Representative.

Specifications

Specifications

12 x 16 x 8 in. (30.5 x 40.5 x 20.95cm)
Welded corners and bottom
0.48cm
Polypropylene
20.95cm
40.5cm
8 in.
12 in.
16 in.
Heavy duty rectangular polypropylene lab tray
30.5cm
No
Documents