missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Heavy Duty Polypropylene Lab Trays

Fabricated with 3/16 in. thick polypropylene.

Manufacturer:  Fisherbrand™ 107334

 View more versions of this product

Catalog No. 15-236-1D



Description

Description

Fabricated with 3/16 in. thick polypropylene.

  • Rectangular with welded corners and bottom
  • Can be customized with spigotes and covers
  • Won't rust

Order Info

Additional spill tray sizes available. To order, contact your Fisher Scientific Customer Service Representative.

Specifications

Specifications

22 x 26 x 4 in. (55.8 x 66 x 10cm)
Welded corners and bottom
0.93cm
Polypropylene
10cm
66cm
4 in.
22 in.
26 in.
Heavy duty rectangular polypropylene lab tray
55.8cm
No
Documents