missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Heavy Duty Polypropylene Lab Trays

Fabricated with 3/16 in. thick polypropylene.

Manufacturer:  Fisherbrand™ 107314

 View more versions of this product

Catalog No. 15-236-1B



Description

Description

  • Rectangular with welded corners and bottom
  • Can be customized with spigotes and covers
  • Won't rust

Order Info

Additional spill tray sizes available. To order, contact your Fisher Scientific Customer Service Representative.

Specifications

Specifications

15 x 20 x 3 in. (38 x 51 x 7.6cm)
Welded corners and bottom
0.48cm
Polypropylene
7.6cm
51cm
3 in.
15 in.
20 in.
Heavy duty rectangular polypropylene lab tray
38cm
No
Documents