missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Heavy Duty HDPE Lab Trays

Fabricated with 1/4 in. thich high density polyethylene.

Manufacturer:  Fisherbrand™ 1088747

 View more versions of this product

Catalog No. 15-236-2D



Description

Description

Fabricated with 1/4 in. thich high density polyethylene.

  • Rectangular with welded corners and bottom
  • Can be customized with spigots and covers
  • Won't rust

Order Info

Additional spill containment tray sizes available. To order, contact your Fisher Scientific Customer Service Representative.

Specifications

Specifications

12 x 16 x 8 in. (30.5 x 40.5 x 20.32cm)
Welded corners and bottom
0.48cm
Polyethylene
20.32cm
40.5cm
8 in.
12 in.
16 in.
Heavy duty rectangular HDPE lab tray
30.5cm
No
Documents