Fisherbrand™ Headspace Vials ND20/ND18
Fisherbrand Headspace Vials ND20/ND18 are offered with a rounded bottom, and are specifically designed to withstand high internal pressure.
Manufacturer:  Fisherbrand™ 13622133
 View more versions of this product
Catalog No. 13-622-133
Specifications
Specifications
PerkinElmer (not suitable for TurboMatrix™ 110)
22.5mm
Each
Headspace Vial
75.5mm
20mL
0.88 in.
100/PP box
Round
Amber
Glass
Documents