missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Headspace 20mm Crimp Seals with Septa

Fisherbrand headspace 20mm Crimp Seals with Septa are quick, convenient, and minimize contamination.

Manufacturer:  Fisherbrand™ 13622131

 View more versions of this product

Catalog No. 13-622-131



Description

Description

  • Seals with pre-inserted septa are convenient and minimize contamination due to handling
  • Use magnetic seals with CTC/Leap™ Technologies, Gerstel™ and other magnetic transport autosamplers
  • PurePak™ PTFE/Silicone seals are packed in glass jars to maintain purity up to the point of use
  • Molded polyethylene SepCap™ is suitable for short-term storage or where repeated access to the vial is required

Specifications

Specifications

Aluminum
Each
0.8cm
Crimp Seal
Crimp
0.31 in.
20mm
Tan, White
Documents