Please login to your online account to display your discounted pricing
Fisherbrand™ HDPE Rectangular Carboys
Carboys Ideal for storing and dispensing lab water and reagents.
Manufacturer: fisherbrand™  N3022110020
 View more versions of this product
Catalog No. 03-007-641
  • / Case
Description & Specifications
Specifications
Capacity (English) 5.28 gal.
Capacity 5.28 gal., 9L
Certifications/Compliance FDA, USP VI
Capacity (Metric) 9L
Material HDPE
Autoclavable No
For Use With (Application) Ideal for storing and dispensing lab water and reagents
Mouth Style Yes
Shape Rectangular
Product Type Carboy without spigot