Please login to your online account to display your discounted pricing
Fisherbrand™ HDPE Rectangular Carboys
Carboys Ideal for storing and dispensing lab water and reagents.
$112.67 - $173.07
Specifications
Certifications/Compliance FDA, USP VI
Autoclavable No
Material HDPE
For Use With (Application) Ideal for storing and dispensing lab water and reagents
Mouth Style Yes
View More Specs
Products
Catalog Number Mfr. No. Capacity Product Type Price Quantity    
03-007-642
fisherbrand™
N3022110050
5.28 gal.; 20L Carboy without spigot Case for $132.10
03-007-641
fisherbrand™
N3022110020
5.28 gal., 9L Carboy without spigot Case for $112.67
03-007-643
fisherbrand™
N3023200020
2.38 gal.; 9L Carboy with spigot Case for $136.60
03-007-644
fisherbrand™
N3023200050
5.28 gal.., 20L Carboy with spigot Case for $173.07
Description & Specifications
Specifications
Certifications/Compliance FDA, USP VI
Autoclavable No
Material HDPE
For Use With (Application) Ideal for storing and dispensing lab water and reagents
Mouth Style Yes
Shape Rectangular
Ideal for storing and dispensing lab water and reagents
  • Durable construction for long service life
  • Molded graduations for 1 gallon and 51 increments
  • Choice of materials for general use or autoclavability
  • Space-saving rectangular design
  • Wide mouth closure allows for easy filling and cleaning access
  • Carboys with spigots include a hose barb insert to accommodate -+ to 5/16-in. ID tubing.
  • Spigot spout without the adapter fits 5/8-in. ID tubing
  • Materials meet FDA requirements for food use and USP VI criteria (non-toxic)