Please login to your online account to display your discounted pricing
Fisherbrand™ HDPE Rectangular Carboys
Carboys Ideal for storing and dispensing lab water and reagents.
Manufacturer: fisherbrand™  N3022110050
 View more versions of this product
Catalog No. 03-007-642
  • / Case
Description & Specifications
Specifications
Capacity (English) 5.28 gal.
Certifications/Compliance FDA, USP VI
Capacity (Metric) 20L
Material HDPE
Autoclavable No
Capacity 5.28 gal.; 20L
For Use With (Application) Ideal for storing and dispensing lab water and reagents
Mouth Style Yes
Shape Rectangular
Product Type Carboy without spigot