Please login to your online account to display your discounted pricing
Fisherbrand™ HDPE Jugs
Made of high density polyethylene
$181.57 - $181.57
Specifications
Closure Liner Foam Polyethylene
Closure Material Polypropylene
Closure Size 38-400
Closure Type Screw
Material HDPE
View More Specs
Includes:  Foamed PE-lined caps
Products
Catalog Number Mfr. No. Capacity Shape Price Quantity    
02-912-115
fisherbrand™
W014387
67.62 oz., 2000mL Round Case of 40 Each for $181.57
Description & Specifications
Specifications
Closure Liner Foam Polyethylene
Closure Material Polypropylene
Closure Size 38-400
Closure Type Screw
Material HDPE
Mouth Narrow
  • Narrow mouth is ideal for liquids
  • Available as round- or square-shaped
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.