Please login to your online account to display your discounted pricing

Fisherbrand™ Hazardous Chemical Dispensers


Bottle-top dispensers for use with hazardous and corrosive chemicals

Manufacturer: fisherbrand™  3103740001

 View more versions of this product

Catalog No. 03-405-43

  • / Each
 

Description & Specifications

Specifications

Width (Metric) 50mm
Size (English) 1.77 in.
Height (English) 15 in.
Product Type Chemical Bottle Pourer
Width (English) 1.97 in.
Material PTFE
Size (Metric) 45mm
Height (Metric) 381mm