Fisherbrand™ Hard Surface Adhesive Mats
Ideal for maintaining a critical clean environment
$36.54 - $38.07
Specifications
Color Blue
Dimensions (L x W) 91 x 46cm (36 x 18 in.)
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Application) Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Price Quantity  
19-181-516 Fisherbrand™
FBM07
High tack Case of 4 Each for $38.07
19-181-519 Fisherbrand™
FBM10
Moderate tack Case of 4 Each for $36.54
Specifications
Description
  • Ideal for use in electronic, pharmaceutical, laboratory and food industry cleanrooms
  • When top sheet becomes soiled, peel it off to expose fresh, clean surface
  • Reorder notification on 18th sheet
  • Blue sheets on blue mat
  • Highly effective in dust removal and easy to use
  • Moderate Tack for standard clean needs
  • High Tack for critical environment users
  • Each mat has top and bottom cover sheets
Order Info
30 sheets per mat.
Description & Specifications
Specifications
Blue
91 x 46cm (36 x 18 in.)