Fisherbrand™ Hanging Drop Slides
For examining living microorganisms such as bacteria and yeast in hanging drops
Manufacturer:  Fisherbrand™ ER1518
 View more versions of this product
Catalog No. 12-560A
Specifications Description & Specifications
Specifications
18mm
1
0.7 in.
Examining living microorganisms such as bacteria and yeast in hanging drops
1.4 to 1.6mm
Hanging Drop Slide