Please login to your online account to display your discounted pricing
Fisherbrand™ Graduated Valumetric™ Bottles with Polyvinyl-Lined Caps
Store a variety of liquids and solids—dry powders, specimens and soil samples
$183.65 - $183.65
Specifications
Closure Liner Poly/Vinyl
Closure Material Polypropylene
Closure Type screw cap
Color Clear
Material Soda Lime Glass
View More Specs
Products
Catalog Number Mfr. No. Capacity Closure Size Quantity Price Quantity    
02-911-457
fisherbrand™
W014154
16 oz., 500mL 70-400 24/PK. Case of 24 Each for $183.65
Description & Specifications
Specifications
Closure Liner Poly/Vinyl
Closure Material Polypropylene
Closure Type screw cap
Color Clear
Material Soda Lime Glass
  • Clear, USP Type III soda-lime glass for easy viewing of contents
  • Printed graduations show milliliters and fluid ounces
  • Wide mouth for easy filling
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.