Fisherbrand™ Glass Tooled-Neck Vials with Polyethylene Closures
General purpose
Manufacturer:  Fisherbrand™ KFS60975D3
 View more versions of this product
Catalog No. 03-339-27C
Specifications Description & Specifications
Specifications
Closed bottom
Clear
Borosilicate Glass
11.1mL
Polyethylene
3 dr.
21mm
3 dr.
Sample Vial
50mm
For general purpose