Please login to your online account to display your discounted pricing

Fisherbrand™ Glass Tooled-Neck Vials with Polyethylene Closures


General purpose

Manufacturer: fisherbrand™  KFS60975D2

 View more versions of this product

Catalog No. 03-339-27B

  • / Pack of 144 Each
  • / Case of 10 Pack
 

Description & Specifications

Specifications

Closure Type Open bottom
Diameter (Metric) Outside 19mm
Color Clear
Capacity (English) 2 dr.
Material Borosilicate Glass
Type Sample Vial
Volume (Metric) 7.4mL
Height (Metric) 48mm
Material Closure Polyethylene
For Use With (Application) For general purpose
Volume (English) 2 dr.