Fisherbrand™ 51 Expansion Short Style Glass Shell Vials without Closures
Short-style shell vials are ideal for storing dry products.
Manufacturer:  Fisherbrand™ KFS609313
 View more versions of this product
Catalog No. 03-339-30F
Description
Description
Short-style shell vials are ideal for storing dry products.
  • Plain top design
  • Lab pack quantities
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Specifications
Specifications
Plain
Ideal for storing dry products
65mm
Shell Vial
Clear
Borosilicate Glass
11 mL
Yes
65mm
144
3 dr.
19 mm
Non-sterile