missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Precleaned with Certification Glass EPA Vials

Packaged in partitioned trays

Supplier:  Fisherbrand™ 02912376

 View more versions of this product

Catalog No. 02-912-376



Description

Description

Glass vials for collection of water and soil samples for Volatile Organic Analysis.

  • Borosilicate glass Clear vials for general analysis; amber vials for light-sensitive materials
  • GPI thread, 24-400

Specifications

Specifications

1.1 in.
28 mm
40 mL
3.74 in.
10 dr.
Polypropylene
95 mm
EPA
Clear
Borosilicate Glass
For general analysis
0.125 in. PTFE/silicone septa, open-top polypropylene screw caps
40 mL
Screw Cap
3.17 mm
Vial
SDS
Documents

Documents

Promotions

Promotions