Fisherbrand™ Glass EPA Vials
Packaged in partitioned trays
Manufacturer:  Fisherbrand™ 02912377
 View more versions of this product
Catalog No. 02-912-377
Specifications
Description
Vial, EPA; Fisherbrand; Precleaned w/certification; Amber for light-sensitive materials; Borosilicate glass; Includes 0.125 in. PTFE/silicone septa; Open-top PP screw caps; Capacity: 40mL; O.D. x H: 28 x 95mm; GPI thread: 24 to 400
  • Borosilicate glass Clear vials for general analysis; amber vials for light-sensitive materials
  • GPI thread, 24-400
Description & Specifications
Specifications
EPA Vial with Certification
Amber
95mm
1.35 oz.
28mm
1.1 in.
0.125 in. PTFE/silicone septa, open-top polypropylene screw caps
Screw
40.0mL
40mL
3.74 in.
EPA
Borosilicate Glass
For light sensitive materials
Screw
Polypropylene
10 dr.