Please login to your online account to display your discounted pricing

Fisherbrand™ General-Purpose PTFE Forceps


For use up to 260°C (500°F)

Manufacturer: fisherbrand™  3165040002

 View more versions of this product

Catalog No. 02-617-160

  • / Each
 

Description & Specifications

Specifications

Material PTFE
Length (Metric) 19.68cm
Product Type Tweezer Forceps
Tip Style Square
Quantity (No. per Case) 1/Each
Length (English) 7.75 in.