Please login to your online account to display your discounted pricing

Fisherbrand™ General-Purpose PTFE Forceps


For use up to 260°C (500°F)

Manufacturer: fisherbrand™  3165040001

 View more versions of this product

Catalog No. 02-617-159

  • / Each
 

Description & Specifications

Specifications

Length (Metric) 15.24cm
Length (English) 6 in.
Product Type Tweezer Forceps
Quantity (No. per Case) 1/Each
Tip Style Square
Material PTFE