missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ General Purpose Liquid-in-Glass Partial Immersion Thermometers

Fisherbrand™ General Purpose Liquid-in-Glass Partial Immersion Thermometers are ideal for use in chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries, and distilleries.

Manufacturer:  Hb Instrument B607001020

 View more versions of this product

Catalog No. 13-201-649


 

Specifications Description & Specifications

Specifications

2°F
Chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries and distilleries
Partial
355mm
2
Not Applicable
ISO/IEC 17025:2005, A2LA, NIST
76mm
14 in.
White Back Glass
20° to 500°F
Blue Organic