Please login to your online account to display your discounted pricing
Fisherbrand™ General Protection Disposable SMS Pink Lab Coats
Single-use lab coats provide high durability and splash/dust protection
$54.43 - $54.43
Specifications
Collar Type Standard
Cuff Style Knit
Closure Type Snap
No. of Pockets 3
Latex Free Yes
View More Specs
Products
Catalog Number Mfr. No. Length (English) Size Price Quantity    
17100849
fisherbrand™
17100849
35 in. Small Case of 30 Each for $54.43
Description & Specifications
Specifications
Collar Type Standard
Cuff Style Knit
Closure Type Snap
No. of Pockets 3
Latex Free Yes
Color Pink
Material SMS (Spunbound Melt-Blown Synthetic)
Disposable Yes
Product Type General SMS protection; Three pockets
Roomy pockets for securly storing items like pens or small instruments
Latex-free, three-layer SMS construction
Packed 10 per pack; 30 per case
Excellent particle retention and water vapor transmission
Available in a variety of colors
Knit cuffs hold sleeves in place; standard collar
Three pockets and a five-snap closure
Fabric weight: 50g/m
Note: Other materials, colors and options are available. See Fisherbrand Basic Protection Disposable Polypropylene Lab Coats with Elastic Cuffs, and Fisherbrand Basic Protection Disposable Lab Coats with Pockets. SMS coats are also available in White, Medical Blue, Cranberry, Teal, Purple and Blueberry.