Please login to your online account to display your discounted pricing

Fisherbrand™ Gel-Loading Tips, 1-200μL


An ideal choice for accurately and easily loading wells in gel electrophoresis

Manufacturer: fisherbrand™  H01096RSFIS

 View more versions of this product

Catalog No. 02-707-139

  • / Pack of 960 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Product Type Hinged rack, sterile, 10 × 96
Material Polypropylene
Length (English) 2.75 in.
Length (Metric) 6.98cm
Sterility Sterile
Tip Style Gel, Round
Shape Round
Quantity 960 Pack
Certifications/Compliance RNase, DNase-free
For Use With (Application) For DNA sequencing or loading electrophoresis gels
Volume (Metric) 1 to 200μL
Color Natural