missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Gel-Loading Tips, 1-200μL

An ideal choice for accurately and easily loading wells in gel electrophoresis

Manufacturer:  Fisherbrand™ 010R204SFIS

 View more versions of this product

Catalog No. 02-707-81




Description

Description

An ideal choice for accurately and easily loading wells in gel electrophoresis

  • Round gel-loading tip
  • Easily load gels 0.6mm
  • Applications include Agarose DNA gels, 2-D and SDS PAGE, Capillary gels and Sequencing
  • Certified by independent laboratory

Specifications

Specifications

Pk. of 1020
1 to 200μL
Natural
PP (Polypropylene)
6.98cm
For DNA sequencing or loading electrophoresis gels
Gel Loading
Sterile
Round
2.75 in.
DNase, RNase Free
Documents