Please login to your online account to display your discounted pricing

Fisherbrand™ Gel-Loading Tips, 0.5-10μL


An ideal choice for accurately and easily loading wells in gel electrophoresis

Manufacturer: fisherbrand™  124R204SFIS

 View more versions of this product

Catalog No. 02-707-89

  • / Pack of 1020 Each
  • / Case of 10 Pack
 

Description & Specifications

Specifications

Product Type Gel Loading Tip; Sterile
Material Polypropylene
Length (English) 2.31 in.
Length (Metric) 5.86cm
Sterility Sterile
Color Natural
Tip Style Gel, Round
Shape Round
Quantity 1020 Pack
Certifications/Compliance RNase, DNase-free
For Use With (Application) For DNA sequencing or loading electrophoresis gels
Volume (Metric) 0.5 to 10μL