Please login to your online account to display your discounted pricing

Fisherbrand™ Gel-Loading Tips, 0.1μL


An ideal choice for accurately and easily loading wells in gel electrophoresis

Manufacturer: fisherbrand™  103R204SFIS

 View more versions of this product

Catalog No. 02-707-84

  • / Pack of 1020 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Product Type Gel Loading Tip
Volume (Metric) 0.1μL
Sterility Sterile
Color Natural
Tip Style Gel, Round
Shape Round
Material Polypropylene
Quantity 1020 Pack
Length (English) 2.25 in.
Length (Metric) 5.7cm
Certifications/Compliance RNase, DNase-free
For Use With (Application) For DNA sequencing or loading electrophoresis gels