Please login to your online account to display your discounted pricing

Fisherbrand™ Gel-Loading Tips, 0.1μL


An ideal choice for accurately and easily loading wells in gel electrophoresis

Manufacturer: fisherbrand™  103R204FIS

 View more versions of this product

Catalog No. 02-707-83

  • / Pack of 1020 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Product Type Gel Loading Tip
Material Polypropylene
Length (English) 2.25 in.
Length (Metric) 5.7cm
Sterility Sterile
Tip Style Gel, Round
Shape Round
Quantity 1020 Pack
Certifications/Compliance RNase, DNase-free
For Use With (Application) For DNA sequencing or loading electrophoresis gels
Volume (Metric) 0.1μL
Color Natural