Please login to your online account to display your discounted pricing

Fisherbrand™ Gel-Loading Aerosol-Barrier Tips


Help minimize liquid retention

Manufacturer: fisherbrand™  FA01020FIS

 View more versions of this product

Catalog No. 02-707-169

  • / Pack of 1000 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Product Type Gel Loading Filter Pipetter Tip
Volume (Metric) 20μL
Color Natural
Tip Style Aerosol Barrier
Material Polypropylene
Quantity 1000 Pack
Length (English) 2.7 in.
Length (Metric) 7cm
For Use With (Application) Help minimize liquid retention