Please login to your online account to display your discounted pricing

Fisherbrand™ Gel-Loading Aerosol-Barrier Tips


Help minimize liquid retention

Manufacturer: fisherbrand™  F010R204S100FIS

 View more versions of this product

Catalog No. 02-707-174

  • / Pack of 1020 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Product Type Gel Loading Filter Pipetter Tip
Volume (Metric) 100μL
Color Natural
Tip Style Aerosol Barrier
Material Polypropylene
Quantity 1020 Pack
Length (English) 2.7 in.
Length (Metric) 7cm
For Use With (Application) Help minimize liquid retention