Please login to your online account to display your discounted pricing

Fisherbrand™ Gel-Loading Aerosol-Barrier Tips


Help minimize liquid retention

Manufacturer: fisherbrand™  FA010100FIS

 View more versions of this product

Catalog No. 02-707-172

  • / Pack of 1000 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Length (Metric) 7cm
Length (English) 2.7 in.
Material Polypropylene
Product Type Gel Loading Filter Pipetter Tip
Volume (Metric) 100μL
Tip Style Aerosol Barrier
Quantity 1000 Pack
Color Natural
For Use With (Application) Help minimize liquid retention