Fisherbrand™ Gas Diffusing Stones
Fast gas diffusion into liquids
$224.00 - $227.07
Specifications
Material Crystalline alumina
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
11-139B Fisher Scientific
ME46944C
Cylindrical 0.875 in. dia. Gas diffusing stones Each for $224.00
11-139A Fisher Scientific
ME46239C
Spherical 1 in. dia. Gas diffusing stones Each for $227.07
Specifications
Description
  • Made of porous, fused crystalline alumina grains
  • Average pore size 60μm
  • Particle retention 25μm With cadmium-plated nipples for 0.187 in. I.D. (4.8mm) tubing
Description & Specifications
Specifications
Crystalline alumina