Fisherbrand™ White Polypropylene PE Lined Foam Cap
Offer good chemical resistance. Fisherbrand™ White Polypropylene PE Foam Caps are known for good impact strength and pliability.
Manufacturer:  Fisherbrand™ 263829
 View more versions of this product
Catalog No. 02-912-095
Description
Description
Resistant against acids, alcohols, alkalis, aqueous products, cosmetics, household oils and solvents
Specifications
Specifications
Plastic
White, White
Polypropylene, Polypropylene
PE Foam Lined Cap
28-400
Screw cap
Foam Polyethylene
Screw cap
4300/Cs.
28/400
Round
Documents