missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Foamed PE-Lined Polypropylene White Caps

For general purpose use. Fisherbrand™ Foamed PE-Lined Polypropylene White Caps offer good impact strength and pliability.

$185.13 - $185.13

Specifications

Certifications/Compliance ISO 9001
Closure Liner Polyethylene Foam
Color White
For Use With (Equipment) Bottle
View More Specs

Products 1
Catalog Number Mfr. No. Neck Finish Material No. per Case Price Quantity  
Catalog Number Mfr. No. Neck Finish Material No. per Case Price Quantity  
02-912-099
View Documents
Fisherbrand™
263834L
53-400 Polypropylene 500 Cs.
Case of 500 Each for $185.13
 
Description

Description

  • Liner molds itself around the sealing areas virtually eliminating leakage
  • Chemical resistant including acids, alcohols, alkalis, aqueous products, cosmetics, household oils and solvents
  • Not autoclavable
 
Specifications

Specifications

ISO 9001
Polyethylene Foam
White
Bottle
SDS
Product Certifications