Fisherbrand™ Foamed PE-Lined Polypropylene White Caps
For general purpose use. Fisherbrand™ Foamed PE-Lined Polypropylene White Caps offer good impact strength and pliability.
Manufacturer:  Fisherbrand™ 263832L
 View more versions of this product
Catalog No. 02-912-097
Specifications
Description
Cap, Plastic; Fisherbrand; White; Foamed PE-lined; General purpose; 38mm-400 screw cap size; 500/cs.
  • Liner molds itself around the sealing areas virtually eliminating leakage
  • Chemical resistant including acids, alcohols, alkalis, aqueous products, cosmetics, household oils and solvents
Description & Specifications
Specifications
ISO 9001
White
500 Cs.
500EA/Cs.
Bottle
38-400
Foam Polyethylene
Polypropylene
Packed in 5 bags of 100
Foamed PE Lined Cap
F217
Round