Please login to your online account to display your discounted pricing

Fisherbrand™ Foamed PE-Lined Polypropylene White Caps


For general purpose use. Fisherbrand™ Foamed PE-Lined Polypropylene White Caps offer good impact strength and pliability.

Manufacturer: fisherbrand™  263829L

 View more versions of this product

Catalog No. 02-911-809

  • / Case of 500 Each
 

Description & Specifications

Specifications

For Use With (Equipment) Bottle
Packaging Packed in 5 bags of 100
Neck Finish 28-400
Shape Round
Liner Material F217
Material Polypropylene
Quantity 500EA/Cs.
Color White
Certifications/Compliance ISO 9001
Closure Liner Foam Polyethylene
No. per Case 500 Cs.

  • Liner molds itself around the sealing areas virtually eliminating leakage
  • Chemical resistant including acids, alcohols, alkalis, aqueous products, cosmetics, household oils and solvents