missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Mini-tip Foam Swab, Black Polypropylene Handle

Flexible mini-tip foam applicator with black plastic handle. Tip is comprised of an open-cell polyurethane foam. Ideal for a variety of cleaning applications.

Manufacturer:  Fisherbrand™ 1805PFRECTFISHER

 View more versions of this product

Catalog No. 14-960-3L




Description

Description

Flexible mini-tip foam applicator with black plastic handle. Tip is comprised of an open-cell polyurethane foam. Ideal for a variety of cleaning applications.

Specifications

Specifications

ISO 9001:2008, ISO 13485:2003, FDA GMP's
0.687 in.
1.745 cm
Medical
5.00 in.
12.7 cm
13.1cm
0.375 in.
Rectangular
Swab
0.245 in.
0.622 cm
Coating rechargeable copier cartridges, applying solutions or solvents, general cleaning
Polypropylene
0.89 in.
2.2cm
Non-sterile
Polyurethane Foam
5.17 in.
Documents