Fisherbrand™ Foam Culture Tube Racks
Resilient wells grip tubes securely
Manufacturer:  Fisherbrand™ BIP0011
 View more versions of this product
Catalog No. 05-664-15B
Description
Description
  • Made from chemical- and solvent-resistant polyester foam
  • Five rows of wells and 10 columns per rack
  • Float in water bath with full load of tubes and samples
  • Flame retardant
Order Info
Weight: Less than 2 oz. (56.7g)
Specifications
Specifications
Not Autoclavable
16 mm Tubes
50
Culture tubes
Foam
Tube Rack
Documents