Please login to your online account to display your discounted pricing

Fisherbrand™ Fluted Qualitative Filter Paper Circles


Cellulose fiber papers

Manufacturer: fisherbrand™  WHT0979014F

 View more versions of this product

Catalog No. 09-790-14F

  • / Pack of 100 Each
  • / Case of 20 Pack
 

Description & Specifications

Specifications

For Use With (Application) Use for coarse particles and gelatinous precipitates.
Porosity Coarse
Particle Retention 25μm
Flow Rate Fast
Format Circle
Diameter (Metric) 18.5cm
Grade P8 Fluted
Accessories

Accessories for Catalog Number: 0979014F

Item Mfr. No. Item Description Price Per Unit Quantity