Please login to your online account to display your discounted pricing

Fisherbrand™ Fluted Qualitative Filter Paper Circles


Cellulose fiber papers

Manufacturer: fisherbrand™  WHT0979014E

 View more versions of this product

Catalog No. 09-790-14E

  • / Pack of 100 Each
 

Description & Specifications

Specifications

Format Circle
Flow Rate Fast
Particle Retention 25μm
Diameter (Metric) 15cm
Grade P8 Fluted
For Use With (Application) Use for coarse particles and gelatinous precipitates.
Porosity Coarse
Accessories

Accessories for Catalog Number: 0979014E

Item Mfr. No. Item Description Price Per Unit Quantity  
09-753-50
Fisherbrand™ Filter Forceps
fisherbrand™
SDI25510
Easy, effortless handling of membranes and glass fiber filters Each for $11.07