missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Fluted Qualitative Filter Paper Circles

Cellulose fiber papers

$55.91 - $141.84

Specifications

Format Circle
Grade P8 Fluted
Porosity Coarse
Flow Rate Fast
For Use With (Application) Use for coarse particles and gelatinous precipitates.
View More Specs

Products 4
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
09-790-14D View Documents Fisherbrand™
0979014D
12.5cm Pack of 100 Each for $108.29
 
09-790-14E Accessories View Documents Fisherbrand™
0979014E
15cm Pack of 100 Each for $55.91
 
09-790-14F Accessories View Documents Fisherbrand™
0979014F
18.5cm Pack of 100 Each for $122.31
Case of 20 Pack for $2,446.20
 
09-790-14G Accessories View Documents Fisherbrand™
0979014G
24cm Pack of 100 Each for $141.84
 
Description

Description

Cellulose fiber papers

  • For use with conical funnels
  • High wet strength†

Specifications

Specifications

Circle
Coarse
Use for coarse particles and gelatinous precipitates.
P8 Fluted
Fast
25μm